Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CGB8
- Entrez Gene ID
-
94115
- Gene Name
- chorionic gonadotropin beta subunit 8
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 49047638 - 49049111 on Build GRCh38
- Gene Symbol
- CGB3
- Entrez Gene ID
-
1082
- Gene Name
- chorionic gonadotropin beta subunit 3
- Gene Aliases
-
CGB, CGB5, CGB7, CGB8, hCGB
- Chromosome Location
-
Chr.19: 49022869 - 49024375 on Build GRCh38
- Gene Symbol
- CGB5
- Entrez Gene ID
-
93659
- Gene Name
- chorionic gonadotropin beta subunit 5
- Gene Aliases
-
CGB, HCG, hCGB
- Chromosome Location
-
Chr.19: 49043845 - 49045311 on Build GRCh38
View Full Product Details