Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RMST
- Entrez Gene ID
-
196475
- Gene Name
- rhabdomyosarcoma 2 associated transcript (non-protein coding)
- Gene Aliases
-
LINC00054, NCRMS, NCRNA00054
- Chromosome Location
-
Chr.12: 97465021 - 97533766 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF429306.1 |
- |
6 - 7 |
1081 |
91 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 135a-2 |
MIR135A2 |
MIRN135-2, MIRN135A2, mir-135a-2 |
-239.0
|
Sense |
Chr. - 97563812 - 97563911 on Build GRCh38 |
Downstream |
platelet activating factor acetylhydrolase 1b catalytic subunit 2 pseudogene |
LOC643711 |
|
148.4kb
|
Anti-sense |
Chr. - 97713412 - 97756517 on Build GRCh38 |
View Full Product Details