Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RMST
- Entrez Gene ID
-
196475
- Gene Name
- rhabdomyosarcoma 2 associated transcript (non-protein coding)
- Gene Aliases
-
LINC00054, NCRMS, NCRNA00054
- Chromosome Location
-
Chr.12: 97465021 - 97533766 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024037.1 |
|
2 - 3 |
114 |
72 |
GenBank mRNA |
AK056164.1 |
- |
2 - 3 |
118 |
72 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA347802.1 |
- |
2 - 3 |
104 |
|
Chr. 12 - 97465031 - 97494778 [+] on Build GRCh38 |
72 |
GenBankmRNA |
DA746093.1 |
- |
2 - 3 |
115 |
|
Chr. 12 - 97465020 - 97494768 [+] on Build GRCh38 |
72 |
GenBankmRNA |
DA753220.1 |
- |
2 - 3 |
106 |
|
Chr. 12 - 97465029 - 97493985 [+] on Build GRCh38 |
72 |
GenBankmRNA |
DA700233.1 |
- |
2 - 3 |
118 |
|
Chr. 12 - 97465021 - 97494711 [+] on Build GRCh38 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 1251 |
MIR1251 |
MIRN1251, hsa-mir-1251 |
|
Sense |
Chr. - 97491909 - 97491978 on Build GRCh38 |
Downstream |
microRNA 135a-2 |
MIR135A2 |
MIRN135-2, MIRN135A2, mir-135a-2 |
71.2kb
|
Sense |
Chr. - 97563812 - 97563911 on Build GRCh38 |
View Full Product Details