Product Details

Assay Reference Genome
Location

Chr.7:37858220 on build GRCh38
Cytoband
7p14.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
51314
Gene Symbol
NME8
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
Location
Chr.7:37848597-37900401 on build GRCh38
Assay Gene Location
Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NME8 NM_016616.4 NP_057700.3
AF202051.2 AAF20909.2
AF305596.1 AAN04258.1
AK313599.1
BC036816.1 AAH36816.1
GQ472221.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv8074 Chr7:37825738 - 38170485 on Build GRCh38 Loss SFRP4 EPDR1 NME8

View Full Product Details