Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
TFR2 | NM_001206855.1 | NP_001193784.1 | ||
NM_003227.3 | NP_003218.2 | |||
XM_005250553.4 | XP_005250610.1 | |||
XM_017012573.1 | XP_016868062.1 | |||
AF067864.1 | AAD45561.1 | |||
AF085928.1 | ||||
AK000421.1 | BAA91153.1 | |||
BC142630.1 | ||||
BC150192.1 | ||||
BI917390.1 | ||||
BX384750.2 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734888 | Chr7:99975623 - 101290285 on Build GRCh38 | Deletion | PILRA TRIP6 TAF6 MIR4658 ZAN MIR4653 ACHE GAL3ST4 C7orf43 ZNHIT1 UFSP1 LRCH4 MIR93 AP1S1 TFR2 GNB2 TRIM56 STAG3L5P-PVRIG2P-PILRB AGFG2 MOSPD3 MBLAC1 CNPY4 MEPCE SERPINE1 MIR25 COPS6 PCOLCE-AS1 EPO NYAP1 PILRB SLC12A9 AP4M1 MUC17 MOGAT3 MCM7 C7orf61 LOC102724094 STAG3L5P SRRT POP7 GPC2 STAG3 PVRIG2P PPP1R35 PMS2P1 ZCWPW1 PLOD3 SAP25 VGF PVRIG AZGP1P1 LAMTOR4 GATS CLDN15 LOC105375429 GIGYF1 AZGP1 EPHB4 MIR6875 FIS1 ZSCAN21 FBXO24 MUC12 ZNF3 SPDYE3 MIR106B PCOLCE MUC3A ZKSCAN1 TSC22D4 ACTL6B MIR6840 ZASP NAT16 |
nsv951386 | Chr7:100566478 - 100682977 on Build GRCh38 | Deletion | FBXO24 MOSPD3 SAP25 PCOLCE PCOLCE-AS1 ACTL6B LRCH4 LOC105375429 ZASP GIGYF1 TFR2 GNB2 AGFG2 |
nsv831072 | Chr7:100564880 - 100765212 on Build GRCh38 | Loss | FBXO24 MOSPD3 ZAN SAP25 PCOLCE PCOLCE-AS1 EPO ACTL6B LRCH4 LOC105375429 ZASP GIGYF1 TFR2 GNB2 POP7 AGFG2 |