Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STEAP1B
Entrez Gene ID
256227
Gene Name
STEAP family member 1B
Gene Aliases
-
Chromosome Location
Chr.7: 22419444 - 22500282 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164460.1 NP_001157932.1 1 - 2 171 140
NM_207342.2 NP_997225.1 1 - 2 171 140
GenBank mRNA AK308991.1 - 1 - 2 68 140
BC066301.1 - 1 - 2 68 140

View Full Product Details