Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NMRK1
- Entrez Gene ID
-
54981
- Gene Name
- nicotinamide riboside kinase 1
- Gene Aliases
-
C9orf95, NRK1, bA235O14.2
- Chromosome Location
-
Chr.9: 75060577 - 75088319 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_023352.1 |
|
1 - 2 |
211 |
100 |
GenBank mRNA |
AK097144.1 |
- |
1 - 2 |
184 |
100 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BP264460.1 |
- |
1 - 2 |
210 |
|
Chr. 9 - 75077524 - 75088216 [-] on Build GRCh38 |
100 |
GenBankmRNA |
DA938831.1 |
- |
1 - 2 |
184 |
|
Chr. 9 - 75077511 - 75088190 [-] on Build GRCh38 |
100 |
GenBankmRNA |
BI517475.1 |
- |
1 - 2 |
132 |
|
Chr. 9 - 75069045 - 75088138 [-] on Build GRCh38 |
100 |
GenBankmRNA |
HY066527.1 |
- |
1 - 2 |
148 |
|
Chr. 9 - 75078333 - 75088154 [-] on Build GRCh38 |
100 |
GenBankmRNA |
HY060872.1 |
- |
1 - 2 |
148 |
|
Chr. 9 - 75077508 - 75088154 [-] on Build GRCh38 |
100 |
GenBankmRNA |
HY095590.1 |
- |
1 - 2 |
148 |
|
Chr. 9 - 75077559 - 75088154 [-] on Build GRCh38 |
100 |
GenBankmRNA |
HY103955.1 |
- |
1 - 2 |
148 |
|
Chr. 9 - 75078358 - 75088154 [-] on Build GRCh38 |
100 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
carnosine N-methyltransferase 1 |
CARNMT1 |
C9orf41, UPF0586 |
-54.6kb
|
Sense |
Chr. - 74980794 - 75028456 on Build GRCh38 |
Downstream |
osteoclast stimulating factor 1 |
OSTF1 |
OSF, SH3P2, bA235O14.1 |
265.0
|
Anti-sense |
Chr. - 75088480 - 75147201 on Build GRCh38 |
View Full Product Details