The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024035.2 | 8 - 9 | 1107 | 61 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 354A | ZNF354A | EZNF, HEL104, HKL1, KID-1, KID1, TCF17 | -37.8kb | Sense | Chr. - 178711521 - 178730702 on Build GRCh38 |
Downstream | zinc finger protein 354B | ZNF354B | KID2 | 87.3kb | Anti-sense | Chr. - 178859813 - 178901135 on Build GRCh38 |