Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LINC01548
Entrez Gene ID
728409
Gene Name
long intergenic non-protein coding RNA 1548
Gene Aliases
C21orf54
Chromosome Location
Chr.21: 33165470 - 33170236 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024102.1 3 - 4 374 99
GenBank mRNA BC105115.1 - 3 - 4 374 99
BC105117.1 - 3 - 4 374 99
BX116511.1 - 3 - 4 327 99


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA868122.1 - 1 - 2 Chr. 21 - 33165527 - 33169828 [+] on Build GRCh38 99
GenBankmRNA AA400173.1 - 3 - 4 327 Chr. 21 - 33165535 - 33170189 [-] on Build GRCh38 99
GenBankmRNA AA400147.1 - 1 - 2 Chr. 21 - 33165470 - 33169802 [+] on Build GRCh38 99
GenBankmRNA AA934973.1 - 1 - 2 Chr. 21 - 33165528 - 33169823 [+] on Build GRCh38 99


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC101928107 LOC101928107 -41.8kb Sense Chr. - 33111704 - 33123700 on Build GRCh38
Downstream interferon alpha and beta receptor subunit 2 IFNAR2 IFN-R, IFN-alpha-REC, IFNABR, IFNARB, IMD45 61.3kb Anti-sense Chr. - 33229895 - 33264513 on Build GRCh38

View Full Product Details