Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PSMG4
- Entrez Gene ID
-
389362
- Gene Name
- proteasome assembly chaperone 4
- Gene Aliases
-
C6orf86, PAC4, bA506K6.2
- Chromosome Location
-
Chr.6: 3254325 - 3308224 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK096543.1 |
- |
1 - 1 |
2553 |
96 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB325873.1 |
- |
1 |
|
|
Chr. 6 - 3230146 - 3230708 [-] on Build GRCh38 |
96 |
GenBankmRNA |
CK299173.1 |
- |
1 |
247 |
|
Chr. 6 - 3230246 - 3230747 [+] on Build GRCh38 |
96 |
GenBankmRNA |
BX435169.2 |
- |
1 |
|
|
Chr. 6 - 3230108 - 3230631 [-] on Build GRCh38 |
96 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC100507194 |
LOC100507194 |
|
-32.2kb
|
Anti-sense |
Chr. - 3182817 - 3195767 on Build GRCh38 |
View Full Product Details