The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_038865.1 | 1 - 2 | 163 | 70 | |
GenBank mRNA | AK097999.1 | - | 1 - 2 | 164 | 70 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | cactin, spliceosome C complex subunit | CACTIN | C19orf29, NY-REN-24, fSAPc | Anti-sense | Chr. - 3610628 - 3626815 on Build GRCh38 | |
Downstream | phosphatidylinositol-4-phosphate 5-kinase type 1 gamma | PIP5K1C | LCCS3, PIP5K-GAMMA, PIP5K1-gamma, PIP5Kgamma | 18.9kb | Anti-sense | Chr. - 3630181 - 3700492 on Build GRCh38 |