Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HPF1
- Entrez Gene ID
-
54969
- Gene Name
- histone PARylation factor 1
- Gene Aliases
-
C4orf27
- Chromosome Location
-
Chr.4: 169729468 - 169757954 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA959986.1 |
- |
1 |
245 |
|
Chr. 4 - 169731153 - 169731705 [-] on Build GRCh38 |
108 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chloride voltage-gated channel 3 |
CLCN3 |
CLC3, ClC-3 |
-6.3kb
|
Anti-sense |
Chr. - 169620521 - 169723187 on Build GRCh38 |
Downstream |
uncharacterized LOC100506085 |
LOC100506085 |
|
186.1kb
|
Sense |
Chr. - 169917761 - 169975902 on Build GRCh38 |
View Full Product Details