The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011535173.2 | XP_011533475.1 | 9 - 9 | 3348 | 94 |
GenBank mRNA | AK128459.1 | - | 1 - 1 | 2854 | 94 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | StAR related lipid transfer domain containing 13 | STARD13 | ARHGAP37, DLC2, GT650, LINC00464 | -286.4kb | Anti-sense | Chr. - 33103135 - 33676835 on Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 457 | LINC00457 | 468.9kb | Anti-sense | Chr. - 34435450 - 34640685 on Build GRCh38 |