Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HSD52
- Entrez Gene ID
-
729467
- Gene Name
- uncharacterized LOC729467
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 59131936 - 59146807 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027120.1 |
|
3 - 3 |
431 |
81 |
GenBank mRNA |
DB453888.1 |
- |
3 - 3 |
431 |
81 |
DQ141102.1 |
- |
1 - 1 |
51 |
81 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA397748.1 |
- |
3 |
351 |
|
Chr. 1 - 59132466 - 59146727 [-] on Build GRCh38 |
81 |
GenBankmRNA |
HY013413.1 |
- |
3 |
444 |
|
Chr. 1 - 59132484 - 59146820 [-] on Build GRCh38 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 1358 |
LINC01358 |
|
-44.1kb
|
Anti-sense |
Chr. - 59020476 - 59088247 on Build GRCh38 |
Downstream |
FGGY carbohydrate kinase domain containing |
FGGY |
|
163.8kb
|
Anti-sense |
Chr. - 59296405 - 59762730 on Build GRCh38 |
View Full Product Details