The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026597.1 | 3 - 4 | 1517 | 67 | |
GenBank mRNA | AK090954.1 | - | 3 - 4 | 1520 | 67 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | DIRC3 antisense RNA 1 | DIRC3-AS1 | -89.7kb | Anti-sense | Chr. - 217282733 - 217344204 on Build GRCh38 | |
Downstream | tensin 1 | TNS1 | MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS | 321.2kb | Sense | Chr. - 217799789 - 217978853 on Build GRCh38 |