Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PDE6G
- Entrez Gene ID
-
5148
- Gene Name
- phosphodiesterase 6G
- Gene Aliases
-
PDEG, RP57
- Chromosome Location
-
Chr.17: 81650459 - 81663418 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026872.1 |
|
1 - 2 |
87 |
80 |
GenBank mRNA |
BX647779.1 |
- |
1 - 2 |
81 |
80 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BQ637120.1 |
- |
1 - 2 |
75 |
|
Chr. 17 - 81650657 - 81656565 [-] on Build GRCh38 |
80 |
GenBankmRNA |
BX509907.1 |
- |
1 - 2 |
81 |
|
Chr. 17 - 81650780 - 81656571 [-] on Build GRCh38 |
80 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
tetraspanin 10 |
TSPAN10 |
OCSP |
-2.9kb
|
Anti-sense |
Chr. - 81637171 - 81648755 on Build GRCh38 |
Downstream |
oxidoreductase like domain containing 1 |
OXLD1 |
C17orf90 |
8.5kb
|
Sense |
Chr. - 81665036 - 81666637 on Build GRCh38 |
View Full Product Details