Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLYATL3
Entrez Gene ID
389396
Gene Name
glycine-N-acyltransferase like 3
Gene Aliases
C6orf140, bA28H17.2
Chromosome Location
Chr.6: 49499958 - 49528064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010904.1 NP_001010904.1 4 - 5 426 102

View Full Product Details