Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BDNF
Entrez Gene ID
627
Gene Name
brain derived neurotrophic factor
Gene Aliases
ANON2, BULN2
Chromosome Location
Chr.11: 27654893 - 27722058 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143814.1 NP_001137286.1 2 - 3 432 117
NM_001143816.1 NP_001137288.1 1 - 2 830 117
GenBank mRNA AY054406.3 - 1 - 2 120 117
EF689010.1 - 1 - 2 844 117
EF689016.1 - 2 - 3 432 117

View Full Product Details