Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C14orf37
Entrez Gene ID
145407
Gene Name
chromosome 14 open reading frame 37
Gene Aliases
c14_5376
Chromosome Location
Chr.14: 57993536 - 58152305 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017021029.1 XP_016876518.1 8 - 8 13458 73
XM_017021030.1 XP_016876519.1 8 - 8 13598 73
GenBank mRNA BC031314.1 - 1 - 1 851 73


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA W15388.1 - Chr. - - [-] on Build 73
GenBankmRNA AW074703.1 - 1 Chr. 14 - 57993531 - 57993836 [+] on Build GRCh38 73
GenBankmRNA AW236442.1 - 1 Chr. 14 - 57993550 - 57993953 [+] on Build GRCh38 73
GenBankmRNA BE464436.1 - 1 Chr. 14 - 57993547 - 57994091 [+] on Build GRCh38 73
GenBankmRNA DB551414.1 - 1 364 Chr. 14 - 57993547 - 57994044 [-] on Build GRCh38 73
GenBankmRNA AI783629.1 - 1 Chr. 14 - 57993514 - 57993806 [+] on Build GRCh38 73
GenBankmRNA R60313.1 - Chr. - - [-] on Build 73
GenBankmRNA AW449028.1 - 1 Chr. 14 - 57993545 - 57993963 [+] on Build GRCh38 73
GenBankmRNA AA724332.1 - 1 Chr. 14 - 57993550 - 57993936 [+] on Build GRCh38 73
GenBankmRNA AA890668.1 - 1 Chr. 14 - 57993545 - 57993849 [+] on Build GRCh38 73
GenBankmRNA BF059276.1 - 1 Chr. 14 - 57993547 - 57994156 [+] on Build GRCh38 73
GenBankmRNA AA973133.1 - 1 Chr. 14 - 57993546 - 57993883 [+] on Build GRCh38 73
GenBankmRNA AI582600.1 - 1 Chr. 14 - 57993550 - 57994090 [+] on Build GRCh38 73
GenBankmRNA Z38275.1 - Chr. - - [-] on Build 73
GenBankmRNA W23675.1 - Chr. - - [-] on Build 73
GenBankmRNA AI655377.1 - 1 Chr. 14 - 57993545 - 57993965 [+] on Build GRCh38 73
GenBankmRNA BQ005133.1 - 1 Chr. 14 - 57993545 - 57993997 [+] on Build GRCh38 73
GenBankmRNA BX107469.1 - 1 Chr. 14 - 57993545 - 57994035 [+] on Build GRCh38 73
GenBankmRNA DB301604.1 - 1 Chr. 14 - 57993547 - 57994094 [+] on Build GRCh38 73
GenBankmRNA AI263065.1 - 1 Chr. 14 - 57993547 - 57993965 [+] on Build GRCh38 73
GenBankmRNA AW243019.1 - 1 Chr. 14 - 57993545 - 57993923 [+] on Build GRCh38 73
GenBankmRNA F03414.1 - Chr. - - [-] on Build 73
GenBankmRNA AA873868.1 - 1 Chr. 14 - 57993547 - 57993874 [+] on Build GRCh38 73


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream solute carrier family 35 member F4 SLC35F4 C14orf36, c14_5373 -11.3kb Sense Chr. - 57563922 - 57982329 on Build GRCh38
Downstream actin-related protein 10 homolog ACTR10 ACTR11, Arp10, Arp11, HARP11 205.6kb Anti-sense Chr. - 58200115 - 58235636 on Build GRCh38

View Full Product Details