Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK093656.1 |
- |
1 |
942 |
|
Chr. 7 - 23629083 - 23631264 [+] on Build GRCh38 |
98 |
GenBankmRNA |
AI968190.1 |
- |
2 |
|
|
Chr. 7 - 23629808 - 23632488 [-] on Build GRCh38 |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
coiled-coil domain containing 126 |
CCDC126 |
|
|
Sense |
Chr. - 23597333 - 23644708 on Build GRCh38 |
Downstream |
family with sequence similarity 221 member A |
FAM221A |
C7orf46 |
48.2kb
|
Sense |
Chr. - 23680114 - 23703249 on Build GRCh38 |
View Full Product Details