Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CN397641.1 |
- |
1 |
317 |
|
Chr. 1 - 2185446 - 2185871 [-] on Build GRCh38 |
70 |
GenBankmRNA |
BX442582.2 |
- |
1 |
149 |
|
Chr. 1 - 2184698 - 2185703 [-] on Build GRCh38 |
70 |
GenBankmRNA |
CR626695.1 |
- |
1 |
149 |
|
Chr. 1 - 2184468 - 2185703 [-] on Build GRCh38 |
70 |
GenBankmRNA |
DB557772.1 |
- |
1 |
|
|
Chr. 1 - 2185283 - 2185722 [+] on Build GRCh38 |
70 |
GenBankmRNA |
HY138124.1 |
- |
1 |
|
|
Chr. 1 - 2185500 - 2185945 [+] on Build GRCh38 |
70 |
GenBankmRNA |
DB568755.1 |
- |
1 |
|
|
Chr. 1 - 2185209 - 2185722 [+] on Build GRCh38 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC100506504 |
LOC100506504 |
|
-108.0
|
Sense |
Chr. - 2181811 - 2184176 on Build GRCh38 |
Downstream |
uncharacterized LOC105378592 |
LOC105378592 |
|
23.2kb
|
Sense |
Chr. - 2207597 - 2215193 on Build GRCh38 |
View Full Product Details