The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010872.2 | NP_001010872.1 | 5 - 5 | 5668 | 103 |
GenBank mRNA | AK024927.1 | - | 1 - 1 | 1153 | 102 |