Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK097301.1 - 1 953 Chr. 17 - 50722074 - 50724224 [+] on Build GRCh38 71


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ankyrin repeat domain 40 ANKRD40 -14.2kb Anti-sense Chr. - 50693190 - 50707909 on Build GRCh38
Downstream long intergenic non-protein coding RNA 483 LINC00483 C17orf73 36.8kb Anti-sense Chr. - 50761034 - 50767515 on Build GRCh38

View Full Product Details