Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK097846.1 - 1 653 Chr. 3 - 49722825 - 49724617 [+] on Build GRCh38 100


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ring finger protein 123 RNF123 FP1477, KPC1 -1.3kb Sense Chr. - 49688753 - 49721529 on Build GRCh38
Downstream cadherin related family member 4 CDHR4 CDH29, PRO34300 66.1kb Anti-sense Chr. - 49790732 - 49802741 on Build GRCh38

View Full Product Details