Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SDIM1
- Entrez Gene ID
-
100505705
- Gene Name
- stress responsive DNAJB4 interacting membrane protein 1
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK129633.1 |
- |
1 - 1 |
1951 |
96 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105378114 |
LOC105378114 |
|
-117.9kb
|
Sense |
Chr. - 165773201 - 165788288 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 473 |
LINC00473 |
C6orf176, bA142J11.1 |
27.8kb
|
Sense |
Chr. - 165924048 - 165988039 on Build GRCh38 |
View Full Product Details