Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CN265563.1 |
- |
1 |
259 |
|
Chr. 7 - 43616089 - 43616757 [-] on Build GRCh38 |
77 |
GenBankmRNA |
AK125381.1 |
- |
1 |
1450 |
|
Chr. 7 - 43615178 - 43617948 [-] on Build GRCh38 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cytochrome c oxidase assembly factor 1 homolog |
COA1 |
C7orf44, MITRAC15 |
|
Sense |
Chr. - 43608452 - 43729541 on Build GRCh38 |
Downstream |
biliverdin reductase A |
BLVRA |
BLVR, BVR, BVRA |
140.2kb
|
Anti-sense |
Chr. - 43758153 - 43807342 on Build GRCh38 |
View Full Product Details