Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG535762.1 |
- |
|
|
|
Chr. - - [-] on Build |
77 |
GenBankmRNA |
AK126109.1 |
- |
1 |
2548 |
|
Chr. 7 - 4685144 - 4688266 [+] on Build GRCh38 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
forkhead box K1 |
FOXK1 |
FOXK1L |
|
Sense |
Chr. - 4644691 - 4771443 on Build GRCh38 |
Downstream |
small nucleolar RNA, C/D box 13 pseudogene 2 |
SNORD13P2 |
RNU13P2, U13, U13.12A |
1.2kb
|
Sense |
Chr. - 4689429 - 4689633 on Build GRCh38 |
View Full Product Details