Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK126813.1 - 1 2145 Chr. 8 - 486117 - 488512 [-] on Build GRCh38 68


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream testis development related protein TDRP C8orf42, Inm01, TDRP1, TDRP2 Sense Chr. - 489790 - 545781 on Build GRCh38
Downstream glutamate rich 1 ERICH1 HSPC319 126.2kb Sense Chr. - 614737 - 731239 on Build GRCh38

View Full Product Details