Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA HY114352.1 - 1 Chr. 9 - 15469250 - 15469724 [-] on Build GRCh38 74
GenBankmRNA AK026895.1 - 1 444 Chr. 9 - 15469232 - 15470951 [+] on Build GRCh38 74


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream PC4 and SFRS1 interacting protein 1 PSIP1 DFS70, LEDGF, PAIP, PSIP2, p52, p75 Anti-sense Chr. - 15464066 - 15511019 on Build GRCh38
Downstream coiled-coil domain containing 171 CCDC171 C9orf93, bA536D16.1, bA778P13.1 81.9kb Sense Chr. - 15552874 - 16062431 on Build GRCh38

View Full Product Details