Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK126233.1 - 2 1849 Chr. 14 - 106276556 - 106279889 [+] on Build GRCh38 102


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream immunoglobulin heavy variable 3-23 IGHV3-23 DP47, IGHV323, V3-23, VH26 -8.7kb Anti-sense Chr. - 106268606 - 106269061 on Build GRCh38
Downstream long intergenic non-protein coding RNA 226 LINC00226 C14orf97, NCRNA00226 7.8kb Sense Chr. - 106287674 - 106288372 on Build GRCh38

View Full Product Details