Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- JKAMP
- Entrez Gene ID
-
51528
- Gene Name
- JNK1/MAPK8-associated membrane protein
- Gene Aliases
-
C14orf100, CDA06, HSPC213, HSPC327, JAMP
- Chromosome Location
-
Chr.14: 59484443 - 59505407 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK128628.1 |
- |
1 - 1 |
249 |
113 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BI521111.1 |
- |
1 |
224 |
|
Chr. 14 - 59501208 - 59501970 [+] on Build GRCh38 |
113 |
GenBankmRNA |
DB234302.1 |
- |
1 |
249 |
|
Chr. 14 - 59501184 - 59501764 [+] on Build GRCh38 |
113 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
trans-L-3-hydroxyproline dehydratase |
L3HYPDH |
C14orf149 |
-16.7kb
|
Anti-sense |
Chr. - 59465545 - 59484438 on Build GRCh38 |
Downstream |
coiled-coil domain containing 175 |
CCDC175 |
C14orf38, c14_5395 |
362.0
|
Anti-sense |
Chr. - 59504212 - 59577276 on Build GRCh38 |
View Full Product Details