Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC044788.1 - 1 2249 Chr. 14 - 29697199 - 29700932 [-] on Build GRCh38 101


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 1551 LINC01551 C14orf23, c14_5148 -902.4kb Anti-sense Chr. - 28772704 - 28794794 on Build GRCh38
Downstream G2/M-phase specific E3 ubiquitin protein ligase G2E3 KIAA1333, PHF7B 858.2kb Anti-sense Chr. - 30559123 - 30620064 on Build GRCh38

View Full Product Details