Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC071806.1 - 1 52 Chr. 18 - 63151111 - 63151320 [-] on Build GRCh38 77


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream PH domain and leucine rich repeat protein phosphatase 1 PHLPP1 PHLPP, PLEKHE1, PPM3A, SCOP -170.7kb Anti-sense Chr. - 62715439 - 62980443 on Build GRCh38
Downstream 3-ketodihydrosphingosine reductase KDSR DHSR, FVT1, SDR35C1 176.4kb Sense Chr. - 63327736 - 63367273 on Build GRCh38

View Full Product Details