Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC105371712
- Entrez Gene ID
-
105371712
- Gene Name
- uncharacterized LOC105371712
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 28246454 - 28250825 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC082966.1 |
- |
1 - 1 |
1148 |
86 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105371930 |
LOC105371930 |
|
-18.0kb
|
Sense |
Chr. - 28222783 - 28228461 on Build GRCh38 |
Downstream |
leucine-rich repeat-containing protein 37A3 |
LOC101926984 |
|
15.6kb
|
Sense |
Chr. - 28263634 - 28272468 on Build GRCh38 |
View Full Product Details