Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO41
Entrez Gene ID
150726
Gene Name
F-box protein 41
Gene Aliases
FBX41
Chromosome Location
Chr.2: 73254682 - 73284453 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080410.2 NP_001073879.2 10 - 11 2454 64
XM_005264158.3 XP_005264215.1 8 - 9 1373 64
XM_006711950.2 XP_006712013.1 15 - 16 3438 64
XM_011532562.1 XP_011530864.1 11 - 12 2886 64
XM_011532563.2 XP_011530865.1 9 - 10 1811 64
XM_017003421.1 XP_016858910.1 12 - 13 5535 64
XM_017003422.1 XP_016858911.1 15 - 16 5180 64
XM_017003423.1 XP_016858912.1 15 - 16 4561 64
GenBank mRNA AB075820.1 - 10 - 11 2292 64
AK095028.1 - 1 - 2 125 64
AK127503.1 - 7 - 8 935 64
BC112202.1 - 9 - 10 1362 64

View Full Product Details