Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO41
Entrez Gene ID
150726
Gene Name
F-box protein 41
Gene Aliases
FBX41
Chromosome Location
Chr.2: 73254682 - 73284453 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080410.2 NP_001073879.2 4 - 5 1567 73
XM_005264158.3 XP_005264215.1 2 - 3 486 73
XM_006711950.2 XP_006712013.1 9 - 10 2551 73
XM_011532562.1 XP_011530864.1 5 - 6 1999 73
XM_011532563.2 XP_011530865.1 3 - 4 924 73
XM_017003421.1 XP_016858910.1 6 - 7 4648 73
XM_017003422.1 XP_016858911.1 9 - 10 4293 73
XM_017003423.1 XP_016858912.1 9 - 10 3674 73
GenBank mRNA AB075820.1 - 4 - 5 1405 73
BC112202.1 - 3 - 4 475 73

View Full Product Details