Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASNS
Entrez Gene ID
440
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Gene Aliases
ASNSD, TS11
Chromosome Location
Chr.7: 97852117 - 97872542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178075.1 NP_001171546.1 8 - 9 1205 90
NM_001178076.1 NP_001171547.1 6 - 7 971 90
NM_001178077.1 NP_001171548.1 6 - 7 904 90
NM_001673.4 NP_001664.3 8 - 9 1279 90
NM_133436.3 NP_597680.2 8 - 9 1534 90
NM_183356.3 NP_899199.2 9 - 10 1557 90
GenBank mRNA AK000379.1 - 8 - 9 1502 90
AK302189.1 - 8 - 9 1099 90
AK302242.1 - 6 - 7 852 90
AK316224.1 - 6 - 7 906 90
BC008723.2 - 9 - 10 1384 90
BC014621.2 - 8 - 9 1155 90
BT007113.1 - 6 - 7 1028 90
M27396.1 - 8 - 9 1206 90

View Full Product Details