Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIST1H2AA
Entrez Gene ID
221613
Gene Name
histone cluster 1, H2aa
Gene Aliases
H2AA, H2AFR, TH2A, bA317E16.2
Chromosome Location
Chr.6: 25726063 - 25726562 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170745.3 NP_734466.1 1 - 1 53 62
GenBank mRNA BC062211.1 - 1 - 1 57 62

View Full Product Details