The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005185.3 | NP_005176.1 | 1 - 1 | 128 | 91 |
GenBank mRNA | AK313934.1 | - | 1 - 1 | 56 | 91 |
HY060230.1 | - | 1 - 1 | 129 | 91 |