Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOCK11
Entrez Gene ID
139818
Gene Name
dedicator of cytokinesis 11
Gene Aliases
ACG, ZIZ2, bB128O4.1
Chromosome Location
Chr.X: 118495664 - 118686162 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144658.3 NP_653259.3 31 - 32 3531 188
XM_005262368.3 XP_005262425.1 32 - 33 3815 188
XM_005262369.4 XP_005262426.1 32 - 33 3509 188
XM_011531276.2 XP_011529578.1 32 - 33 3450 188
XM_011531278.2 XP_011529580.1 19 - 20 1936 188
GenBank mRNA AK124735.1 - 26 - 27 2954 188
AK125641.1 - 10 - 11 1043 188
AY692226.1 - 31 - 32 3531 188

View Full Product Details