The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144658.3 | NP_653259.3 | 48 - 49 | 5521 | 96 |
XM_005262368.3 | XP_005262425.1 | 49 - 50 | 5805 | 96 | |
XM_005262369.4 | XP_005262426.1 | 49 - 50 | 5499 | 96 | |
XM_011531276.2 | XP_011529578.1 | 49 - 50 | 5440 | 96 | |
XM_011531278.2 | XP_011529580.1 | 36 - 37 | 3926 | 96 | |
GenBank mRNA | AK054649.1 | - | 9 - 10 | 1203 | 96 |
AK056684.1 | - | 18 - 19 | 2035 | 96 | |
AK124735.1 | - | 43 - 44 | 4944 | 96 | |
AK125641.1 | - | 27 - 28 | 3033 | 96 | |
AY692226.1 | - | 48 - 49 | 5521 | 96 | |
BC047713.1 | - | 14 - 15 | 1612 | 96 |