The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144658.3 | NP_653259.3 | 49 - 50 | 5739 | 66 |
XM_005262368.3 | XP_005262425.1 | 50 - 51 | 6023 | 66 | |
XM_005262369.4 | XP_005262426.1 | 50 - 51 | 5717 | 66 | |
XM_011531276.2 | XP_011529578.1 | 50 - 51 | 5658 | 66 | |
XM_011531278.2 | XP_011529580.1 | 37 - 38 | 4144 | 66 | |
GenBank mRNA | AK054649.1 | - | 10 - 11 | 1421 | 66 |
AK056684.1 | - | 19 - 20 | 2253 | 66 | |
AK124735.1 | - | 44 - 45 | 5162 | 66 | |
AK125641.1 | - | 28 - 29 | 3251 | 66 | |
AY692226.1 | - | 49 - 50 | 5739 | 66 | |
BC047713.1 | - | 15 - 16 | 1830 | 66 |