The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172139.2 | NP_742151.2 | 2 - 3 | 259 | 97 |
XM_005258765.4 | XP_005258822.1 | 3 - 4 | 671 | 97 | |
GenBank mRNA | AY129149.1 | - | 2 - 3 | 267 | 97 |
AY336714.1 | - | 2 - 3 | 258 | 97 | |
AY336717.1 | - | 2 - 3 | 258 | 97 | |
BC130314.1 | - | 2 - 3 | 294 | 97 | |
BC130316.1 | - | 2 - 3 | 294 | 97 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172138.1 | NP_742150.1 | 3 - 4 | 319 | 97 |
GenBank mRNA | AY129148.1 | - | 3 - 4 | 319 | 97 |
AY184373.1 | - | 2 - 3 | 255 | 97 | |
AY336715.1 | - | 2 - 3 | 258 | 97 | |
BC113581.1 | - | 3 - 4 | 322 | 97 | |
BC113583.1 | - | 3 - 4 | 322 | 97 | |
DQ126337.1 | - | 3 - 4 | 319 | 97 |