The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002988.3 | NP_002979.1 | 1 - 1 | 67 | 68 |
GenBank mRNA | AB000221.1 | - | 1 - 1 | 59 | 68 |
BC069700.1 | - | 1 - 1 | 50 | 68 | |
BC096124.3 | - | 1 - 1 | 57 | 68 | |
BC096125.3 | - | 1 - 1 | 57 | 68 | |
BC096126.1 | - | 1 - 1 | 57 | 68 | |
BC096127.1 | - | 1 - 1 | 57 | 68 | |
Y13710.1 | - | 1 - 1 | 66 | 68 |