Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AKR7L
Entrez Gene ID
246181
Gene Name
aldo-keto reductase family 7 like (gene/pseudogene)
Gene Aliases
AFAR3, AFB1-AR3, AKR7A4
Chromosome Location
Chr.1: 19265982 - 19274074 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_040288.1 7 - 7 1052 82
NR_040289.1 5 - 5 855 82
GenBank mRNA AJ278012.1 - 6 - 6 838 82
BC035351.1 - 5 - 5 722 82

View Full Product Details