Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- HERC2P7
- Entrez Gene ID
-
100132101
- Gene Name
- hect domain and RLD 2 pseudogene 7
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 22479153 - 22482374 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_036470.1 |
|
2 - 3 |
298 |
69 |
GenBank mRNA |
BC012949.1 |
- |
2 - 3 |
434 |
69 |
- Gene Symbol
- HERC2P11
- Entrez Gene ID
-
100996738
- Gene Name
- hect domain and RLD 2 pseudogene 11
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 28573738 - 28589412 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CA489376.1 |
- |
2 - 3 |
209 |
|
Chr. 15 - 23311422 - 23313710 [+] on Build GRCh38 |
69 |
GenBankmRNA |
BG282592.1 |
- |
|
433 |
|
Chr. - - [-] on Build |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
E3 ubiquitin-protein ligase HERC2-like |
LOC102723534 |
|
-8.1kb
|
Anti-sense |
Chr. - 23288014 - 23303510 on Build GRCh38 |
Downstream |
uncharacterized LOC101929047 |
LOC101929047 |
|
12.8kb
|
Anti-sense |
Chr. - 28589492 - 28618260 on Build GRCh38 |
View Full Product Details