The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190807.2 | NP_001177736.1 | 2 - 3 | 402 | 90 |
GenBank mRNA | CA771478.1 | - | 2 - 3 | 368 | 90 |