The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001745.3 | NP_001736.1 | 1 - 2 | 307 | 89 |
XM_011543653.1 | XP_011541955.1 | 1 - 2 | 860 | 89 | |
GenBank mRNA | AK313156.1 | - | 1 - 2 | 231 | 89 |
BC130325.1 | - | 1 - 2 | 239 | 89 | |
BG720084.1 | - | 1 - 2 | 279 | 89 | |
DA727548.1 | - | 1 - 2 | 316 | 89 | |
U18242.1 | - | 1 - 2 | 202 | 89 |