The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007276.4 | NP_009207.2 | 3 - 4 | 541 | 112 |
NM_016587.3 | NP_057671.2 | 3 - 4 | 313 | 112 | |
XM_005249611.3 | XP_005249668.1 | 3 - 4 | 1048 | 112 | |
GenBank mRNA | AB030905.1 | - | 1 - 2 | 132 | 112 |
AL542267.3 | - | 3 - 4 | 321 | 112 | |
BC000954.1 | - | 3 - 4 | 273 | 112 | |
BX648807.1 | - | 3 - 4 | 269 | 112 | |
CD514780.1 | - | 3 - 4 | 324 | 112 | |
U26312.1 | - | 1 - 2 | 132 | 112 |