The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008493.1 | NP_001008493.1 | 11 - 12 | 1992 | 101 |
XM_005273187.2 | XP_005273244.1 | 12 - 13 | 2032 | 101 | |
XM_011544229.2 | XP_011542531.2 | 12 - 13 | 2508 | 101 | |
XM_017001746.1 | XP_016857235.1 | 13 - 14 | 3267 | 101 | |
XM_017001748.1 | XP_016857237.1 | 11 - 12 | 2451 | 101 | |
XM_017001750.1 | XP_016857239.1 | 12 - 13 | 2409 | 101 | |
XM_017001753.1 | XP_016857242.1 | 14 - 15 | 2071 | 101 | |
XM_017001754.1 | XP_016857243.1 | 13 - 14 | 2015 | 101 | |
XM_017001755.1 | XP_016857244.1 | 13 - 14 | 1758 | 101 | |
XM_017001756.1 | XP_016857245.1 | 11 - 12 | 1689 | 101 | |
XM_017001757.1 | XP_016857246.1 | 11 - 12 | 1592 | 101 | |
GenBank mRNA | AF519769.1 | - | 11 - 12 | 1808 | 101 |
AK127561.1 | - | 5 - 6 | 1123 | 101 | |
BC095481.1 | - | 11 - 12 | 1991 | 101 | |
KM214204.1 | - | 11 - 12 | 1596 | 101 |